Learn More
Akron Biotechnology LLCSupplier Diversity Partner RHU IL-17-F 5UG DFS Item
Manufacturer:  Akron Biotechnology LLCSupplier Diversity Partner AK83120005
rHu IL-17-F is a non-glycosylated protein consisting of two 134 amino acid polypeptide chains that are cysteine linked homodimeric polypeptide chains, with at molecular weight of 30 kDa. 5ug
Catalog No. 50-753-3230