Learn More
Akron Biotechnology LLCSupplier Diversity Partner RHU IL-9 100UG DFS Item
Manufacturer:  Akron Biotechnology LLCSupplier Diversity Partner AK90810100
rHu IL-9 is known to regulate many cell types involved in T- helper type 2 responses classically associated with asthma, including B- and T- lymphocytes, mast cells, eosinophils and epithelial cells.100ug
Catalog No. 50-753-3265