Learn More
Akron Biotechnology LLCSupplier Diversity Partner RHU LEPTIN 5MG DFS Item
Manufacturer:  Akron Biotechnology LLCSupplier Diversity Partner AK82655000
rHu Leptin inhibits food intake and stimulates energy expenditure. Leptin also has thermogenic actions and regulates enzymes of fatty acid oxidation. Severe hereditary obesity in rodents and humans is caused by defects in leptin production.5mg
Catalog No. 50-753-3271