Learn More
Akron Biotechnology LLCSupplier Diversity Partner RHU MCP-3 100UG DFS Item
Manufacturer:  Akron Biotechnology LLCSupplier Diversity Partner AK82610100
rHu MCP-3 is produced in E.Coli is a non-glycosylated, Polypeptide chain containing 76 amino acids and having a molecular mass of 9011 Dalton. 100ug
Catalog No. 50-753-3280