Learn More
Akron Biotechnology LLCSupplier Diversity Partner RHU MIP-1-ALPHA 5UG DFS Item
Manufacturer:  Akron Biotechnology LLCSupplier Diversity Partner AK82500005
rHu MIP-1-alpha is produced in E.Coli is a single, non-glycosylated, polypeptide chain containing 70 amino acids and having a molecular mass of 7820 Dalton. 5ug
Catalog No. 50-753-3287