Learn More
Akron Biotechnology LLCSupplier Diversity Partner RHU MIP-4 100UG DFS Item
Manufacturer:  Akron Biotechnology LLCSupplier Diversity Partner AK82180100
rHu MIP-4 is produced in E.Coli is a single, non-glycosylated, polypeptide chain containing 69 amino acids and having a molecular mass of 7813 Daltons. 100ug
Catalog No. 50-753-3301