Learn More
Akron Biotechnology LLCSupplier Diversity Partner RHU MYOSTATIN 2UG DFS Item
Manufacturer:  Akron Biotechnology LLCSupplier Diversity Partner AK82560002
rHu Myostatin is a member of the transforming growth factor beta superfamily that is essential for proper regulation of skeletal muscle mass. 2ug
Catalog No. 50-753-3302