Learn More
Akron Biotechnology LLCSupplier Diversity Partner RHU NGF-BETA 5UG DFS Item
Manufacturer:  Akron Biotechnology LLCSupplier Diversity Partner AK82790005
rHu NGF-beta acts as a neurotrophic factor through its receptor beta-NGFR, and is involved in the development and sustaining the sensory and sympathetic nervous systems. 5ug
Catalog No. 50-753-3121