Learn More
Akron Biotechnology LLCSupplier Diversity Partner RHU RELM-BETA 25UG DFS Item
Manufacturer:  Akron Biotechnology LLCSupplier Diversity Partner AK90850025
rHu Relm-beta consists of two non-glycosylated polypeptide chains consisting of 89 amino acids with a molecular weight of approximately 19 kDa. 25ug
Catalog No. 50-753-3324