Learn More
Akron Biotechnology LLCSupplier Diversity Partner RHU TNF-ALPHA 10UG DFS Item
Manufacturer:  Akron Biotechnology LLCSupplier Diversity Partner AK83240010
rHu TNF-alpha is secreted by macrophages, monocytes, neutrophils, T-cells, NK- cells following their stimulation by bacterial LPS . Cells expressing CD4 secrete TNF-alpha while CD8 cells secrete little or no TNF-alpha. 10ug
Catalog No. 50-753-3344