Learn More
Akron Biotechnology LLCSupplier Diversity Partner RHU TSLP 10UG DFS Item
Manufacturer:  Akron Biotechnology LLCSupplier Diversity Partner AK90910010
rHu TSLP is produced in several tissues including heart, liver and prostate. TSLP shares similar biological activities to IL-7 including enhancement of STAT3 and STAT5. 10ug
Catalog No. 50-753-3352