Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ RI 1

RAD51 recombinase inhibitor

$109.00 - $465.00

Specifications

CAS 415713-60-9
Formula Weight 361.61g/mol
For Use With (Application) DNA, RNA and Protein Synthesis Inhibitors
Molecular Formula C14H11Cl3N2O3
Storage Requirements Store at 4°C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
61-681-0 View Documents Tocris Bioscience™
6168/10
10mg Each for $109.00
Add to cart
 
61-685-0 View Documents Tocris Bioscience™
6168/50
50mg Each for $465.00
Add to cart
 
Description

Description

RAD51 inhibitor (IC50 is in the range 5 - 30 μM). Covently binds C139 on protein surface. Inhibits homologous recombination. Cell permeable. Sensitizes tumor cells to chemotherapy.
Specifications

Specifications

415713-60-9
DNA, RNA and Protein Synthesis Inhibitors
Store at 4°C
361.61g/mol
C14H11Cl3N2O3

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit