Learn More
United Scientific Supplies RIA Vials Products for Science Education
Available in both polypropylene and polystyrene.
$18.50 - $23.75
Products 2
Catalog Number Mfr. No. Material Price Quantity & Availability  
Catalog Number Mfr. No. Material Price Quantity & Availability  
S04542 Products for Science Education
View Documents
United Scientific Supplies
RVIALPS
Polystyrene Pack of 500 for $23.75
Add to cart
S04541 Products for Science Education
View Documents
United Scientific Supplies
RVIALPP
Polypropylene Pack of 500 for $18.50
Add to cart
Description
Description
  • Polypropylene vials are designed for use with common acids, alkalis and solvents at room temperature
  • Polystyrene vials are designed for use with mild bases and weak acids