Learn More
United Scientific Supplies RIA Vials Products for Science Education
Available in both polypropylene and polystyrene.
Manufacturer:  United Scientific Supplies RVIALPS
 View more versions of this product
Catalog No. S04542
Add to cart
Description
Description
  • Polypropylene vials are designed for use with common acids, alkalis and solvents at room temperature
  • Polystyrene vials are designed for use with mild bases and weak acids
Specifications
Specifications
Common acids, alkalies, and solvents at room temperature
Polystyrene