Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ribavirin, USP, 98.9-101.5%, Spectrum™

Click to view available options
Quantity:
100 g
500 g
1 kg
25 kg
Packaging:
Amber Glass Bottle
Fiber Drum
36791-04-5
C8H12N4O5
244.21
IWUCXVSUMQZMFG-AFCXAGJDSA-N
1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-1H-1,2,4-triazole-3-carboxamide
NC(=O)C1=NN(C=N1)[C@@H]1O[C@H](CO)[C@@H](O)[C@H]1O
36791-04-5
C8H12N4O5
244.21
IWUCXVSUMQZMFG-AFCXAGJDSA-N
1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-1H-1,2,4-triazole-3-carboxamide
NC(=O)C1=NN(C=N1)[C@@H]1O[C@H](CO)[C@@H](O)[C@H]1O

R1052, 36791-04-5, C8H12N4O5

Spectrum™ Ribavirin, USP is an antiviral drug used primarily to treat hepatitis C and viral hemorrhagic fevers. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Packaging Amber Glass Bottle
Ignition Residue 0.0025
Loss on Drying 0.005
Specific Rotation -33.5° to -37.0°
Quantity 100 g
CAS Max % 1
Percent Purity 98.9 to 101.5%
Grade USP