Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ribonuclease A, bovine pancreas, purified, Alfa Aesar™

Used to remove RNA from DNA plasmid preparations and protein samples. Ribonuclease A is used for RNase protection assays, to remove unspecifically bound RNA, analysis of RNA sequences, to hydrolyze RNA contained in protein samples, and the purification of DNA.

$100.80 - $314.00

Specifications

Format Lyophilized Powder
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAJ61996MC View Documents Alfa Aesar
J61996MC
100mg Each for $314.00
Order in Bulk

Request bulk or custom formats

Add to cart
 
AAJ61996MB View Documents Alfa Aesar
J61996MB
25mg Each for $100.80
Order in Bulk

Request bulk or custom formats

Add to cart
 
Specifications

Specifications

Lyophilized Powder
Safety and Handling

Safety and Handling

EINECSNumber : 232-646-6

Recommended Storage : Keep cold

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit