Learn More
Ribonuclease A, bovine pancreas, purified, Alfa Aesar™
Used to remove RNA from DNA plasmid preparations and protein samples. Ribonuclease A is used for RNase protection assays, to remove unspecifically bound RNA, analysis of RNA sequences, to hydrolyze RNA contained in protein samples, and the purification of DNA.
Manufacturer:  Alfa Aesar J61996MB
 View more versions of this product
Catalog No. AAJ61996MB
Order in Bulk
Request bulk or custom formats
Add to cart
Specifications
Specifications
Lyophilized Powder
25mg