Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ribonuclease Inhibitor Mouse anti-Human, PE/Cy5.5, Clone: 1H23, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP130167PECY55
Description
Ribonuclease Inhibitor Monoclonal antibody specifically detects Ribonuclease Inhibitor in Human samples. It is validated for Flow Cytometry.Specifications
Ribonuclease Inhibitor | |
1H23 | |
Flow Cytometry | |
Conjugated, Purified | |
RNH1 | |
Ribonuclease inhibitor 1(RNH1) (7-461aa) purified from E. coli | |
Protein G purified | |
Store at 4°C in the dark. Do not freeze. | |
Monoclonal | |
This antibody is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human RNH1 protein. |
Flow Cytometry | |
PE-Cyanine5.5 | |
PBS with 0.05% Sodium Azide | |
Placental ribonuclease inhibitor, Placental RNase inhibitor, PRI, RAIMGC4569, ribonuclease inhibitor, ribonuclease/angiogenin inhibitor, ribonuclease/angiogenin inhibitor 1MGC18200, RNHMGC54054 | |
Mouse | |
IgG2a Kappa | |
0.1 mL | |
Primary | |
6050.0 | |
Human |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title