Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ribostamycin sulfate, 630μg/mg, Alfa Aesar™

Click to view available options
Quantity:
250mg
1g
53797-35-6
C17H36N4O14S
552.549
MFCD00057496
RTCDDYYZMGGHOE-CZUQXGEQSA-N
ribostamycin sulfate
53486170
(2R,3S,4R,5R,6R)-5-amino-2-(aminomethyl)-6-[(1R,2R,3S,4R,6S)-4,6-diamino-2-[(3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]oxy-3-hydroxycyclohexyl]oxyoxane-3,4-diol;sulfuric acid
C1C(C(C(C(C1N)OC2C(C(C(C(O2)CN)O)O)N)OC3C(C(C(O3)CO)O)O)O)N.OS(=O)(=O)O
53797-35-6
C17H36N4O14S
552.549
MFCD00057496
RTCDDYYZMGGHOE-CZUQXGEQSA-N
ribostamycin sulfate
53486170
(2R,3S,4R,5R,6R)-5-amino-2-(aminomethyl)-6-[(1R,2R,3S,4R,6S)-4,6-diamino-2-[(3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]oxy-3-hydroxycyclohexyl]oxyoxane-3,4-diol;sulfuric acid
C1C(C(C(C(C1N)OC2C(C(C(C(O2)CN)O)O)N)OC3C(C(C(O3)CO)O)O)O)N.OS(=O)(=O)O

Used as a broad spectrum antibiotic targeting both Gram-positive and Gram-negative bacteria. Ribostamycin is a broad-spectrum antimicrobial isolated from Streptomyces ribosifidicus. It is used in pharmacokinetic studies.

Quantity 250mg
Melting Point 175°C to 180°C
Flash Point >110°C
Packaging Plastic bottle
Formula Weight 552.55
Physical Form Powder
Chemical Name or Material Ribostamycin sulfate