Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ribulose-phosphate 3-epimerase Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA5120295

This item is not returnable.
View return policy
Description
Positive test controls include: K-562, HeLa, 293T, Mouse brain, Mouse heart, Mouse thymus, Rat brain.
Ribulose-phosphate 3-epimerase Polyclonal antibody specifically detects Ribulose-phosphate 3-epimerase in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western BlotSpecifications
Ribulose-phosphate 3-epimerase | |
Polyclonal | |
Unconjugated | |
RPE | |
2810429B02Rik; 5730518J08Rik; HUSSY-17; ik:tdsubc_2c8; PE; Ribulose-5-phosphate-3-epimerase; ribulose-5-phosphate-epimerase; Ribulose-phosphate 3-epimerase; Rpe; R-PE; RPE2-1; R-phycoerythrin; wu:fa07h08; wu:fb93d11; wu:fc20b11; xx:tdsubc_2c8 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-228 of human RPE (NP_954699.1). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
2.83 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8VEE0, Q96AT9 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
501157, 6120, 66646 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title