Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RIC3 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548432

Catalog No. PA548432

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Promotes functional expression of homomeric alpha-7 and alpha-8 nicotinic acetylcholine receptors at the cell surface. May also promote functional expression of homomeric serotoninergic 5- HT3 receptors, and of heteromeric acetylcholine receptors alpha- 3/beta-2, alpha-3/beta-4, alpha-4/beta-2 and alpha-4/beta-4.
Specifications

Specifications

RIC3
Polyclonal
PBS with 0.09% sodium azide
Q7Z5B4
Protein RIC-3, Resistant to inhibitor of cholinesterase 3, RIC3
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
79608
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
RIC3
Liquid
RIC3
KLH conjugated synthetic peptide between 313-341 amino acids from the C-terminal region of human RIC3.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit