Learn More
Thermo Scientific™ Richard-Allan Scientific™ Bio-Tite™ Specimen Containers Available on GSA/VA Contract for Federal Government customers only.
Transport, store and retrieve your pathology specimens in a safe and cost-effective manner with Thermo Scientific™ Richard-Allan Scientific™ Bio-Tite™ Specimen Containers.
Manufacturer:  Thermo Scientific™ 599601ZF
 View more versions of this product
Catalog No. 22-110-674
  Check Availability
Add to cart
Specifications
Description
Transport, store and retrieve pathology specimens in a safe and cost effective manner with the Thermo Scientific™ Richard-Allan Scientific™ Specimen Transportation System (STS™). This system includes containers pre-filled with the highest quality 10% Neutral Buffered Formalin (NBF) and Zinc Formalin.
Three unique packaging configurations allow for easy distribution and transport.
53 Series Containers
  • Easy to transport trays ideal for operating rooms and hospitals
  • Clear polystyrene containers have screw-top lids
  • Comply with all updated federal and state OSHA requirements
  • Labeled with highly visible, bright yellow labels with space for patient information for instant identification to minimize health risks
56 Series Containers
  • Patented Thermo Scientific™ Bio-Tite™ closure system is dual-threaded, making it virtually impossible to cross-thread and minimizing leakage
  • Easy to transport trays
  • Comply with all updated federal and state OSHA requirements
  • Labeled with highly visible, bright yellow labels with space for patient information for instant identification to minimize health risks
59 Series Containers
  • Patented Thermo Scientific Bio-Tite closure system is dual-threaded, making it virtually impossible to cross-thread and minimizing leakage
  • Stack pack reduces excess packaging
  • Comply with all updated federal and state OSHA requirements
  • Labeled with highly visible, bright yellow labels with space for patient information for instant identification to minimize health risks
Description & Specifications
Specifications
No
Bio-Tite
Clear
With Bio-Tite™ Cap, Dual-threaded
Polypropylene
960mL
Prefilled Container, Zinc Formalin