Thermo Scientific™ Richard-Allan Scientific™ Bio-Tite™ Specimen Containers
Transport, store and retrieve your pathology specimens in a safe and cost-effective manner with Thermo Scientific™ Richard-Allan Scientific™ Bio-Tite™ Specimen Containers.
Manufacturer: Thermo Scientific™ 53175
 View more versions of this product
Catalog No. 22-110-806
  Check Availability
Add to Cart
    Specifications
    Description Dry Containers
    Sterile No
    Material Polyethylene
    Cap Type Snap
    Capacity (Metric) 175mL
    Transport, store and retrieve pathology specimens in a safe and cost effective manner with the Thermo Scientific™ Richard-Allan Scientific™ Specimen Transportation System (STS™). This system includes containers pre-filled with the highest quality 10% Neutral Buffered Formalin (NBF) and Zinc Formalin.
    Three unique packaging configurations allow for easy distribution and transport.
    53 Series Containers
    • Easy to transport trays ideal for operating rooms and hospitals
    • Clear polystyrene containers have screw-top lids
    • Comply with all updated federal and state OSHA requirements
    • Labeled with highly visible, bright yellow labels with space for patient information for instant identification to minimize health risks
    56 Series Containers
    • Patented Thermo Scientific™ Bio-Tite™ closure system is dual-threaded, making it virtually impossible to cross-thread and minimizing leakage
    • Easy to transport trays
    • Comply with all updated federal and state OSHA requirements
    • Labeled with highly visible, bright yellow labels with space for patient information for instant identification to minimize health risks
    59 Series Containers
    • Patented Thermo Scientific Bio-Tite closure system is dual-threaded, making it virtually impossible to cross-thread and minimizing leakage
    • Stack pack reduces excess packaging
    • Comply with all updated federal and state OSHA requirements
    • Labeled with highly visible, bright yellow labels with space for patient information for instant identification to minimize health risks
  • Description & Specifications
Related Products
Product Content Feedback