Learn More
Thermo Scientific™ Richard-Allan Scientific™ Cal-Rite™ Decalcifying/Fixation Solution
Streamline processes with Thermo Scientific™ Richard-Allan Scientific™ Cal-Rite™ Decalcifying/Fixation Solution, which allows simultaneous decalcification and fixation.
Manufacturer:  Thermo Scientific™ 5501
 View more versions of this product
Catalog No. 14-387-698
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Streamline processes with Thermo Scientific™ Richard-Allan Scientific™ Cal-Rite™ Decalcifying/Fixation Solution, a mixture of formic acid and formaldehyde which allows simultaneous decalcification and fixation.
  • Beneficial for conventional iron staining
  • Preserves nuclear chromatin staining, even after prolonged exposure
  • Eliminates poor hematoxylin and eosin staining caused by over-decalcification and/or exposure to harsh mineral acids.
Specifications
Specifications
Bottle
Bottle
1 gal., Ea.