Learn More
Thermo Scientific™ Richard-Allan Scientific™ Cal-Rite™ Decalcifying/Fixation Solution Available on GSA/VA Contract for Federal Government customers only.
Streamline processes with Thermo Scientific™ Richard-Allan Scientific™ Cal-Rite™ Decalcifying/Fixation Solution, which allows simultaneous decalcification and fixation.
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
22-046339 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
5501
1 gal., Ea. Each for $184.10
N/A
Due to product restrictions, please to purchase or view availability for this product.
14-387-698
View Documents
Thermo Scientific™
5501
1 gal., Ea. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
Streamline processes with Thermo Scientific™ Richard-Allan Scientific™ Cal-Rite™ Decalcifying/Fixation Solution, a mixture of formic acid and formaldehyde which allows simultaneous decalcification and fixation.
  • Beneficial for conventional iron staining
  • Preserves nuclear chromatin staining, even after prolonged exposure
  • Eliminates poor hematoxylin and eosin staining caused by over-decalcification and/or exposure to harsh mineral acids.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.