Learn More
Thermo Scientific™ Richard-Allan Scientific™ Cover Glass Available on GSA/VA Contract for Federal Government customers only.
Reduces defect costs that range from wasted resources to lost time with a consistent, reliable workflow, which includes quality cover glass.
Manufacturer:  Thermo Scientific™ 152430
 View more versions of this product
Catalog No. 22-050-241
  Check Availability
Add to cart
Specifications
Description
Reduces defect costs that range from wasted resources to lost time with a consistent, reliable workflow, which includes quality cover glass. Pre-cleaned Thermo Scientific™ Richard-Allan Scientific™ Cover Glasses are available in assorted sizes and thicknesses.
Ordering Information:
One oz. pack
Description & Specifications
Specifications
24mm
0.16 to 0.19mm
30 x 24mm
30mm
Thickness no.1.5; 24 x 30mm