Thermo Scientific™ Richard-Allan Scientific™ Cover Glass Available on GSA/VA Contract for Federal Government customers only.

Reduces defect costs that range from wasted resources to lost time with a consistent, reliable workflow, which includes quality cover glass.

Manufacturer: Thermo Scientific™ 152460

 View more versions of this product

Catalog No. 22-050-246

  Check Availability
Add to Cart

    Specifications

    Size (Metric) 60 x 24mm
    Item Description Cvr Glass no.1.5 24x60 10 Oz/cs
    Length (Metric) 24mm
    Width (Metric) 60mm
    Thickness (Metric) 0.16 to 0.19mm

    Reduces defect costs that range from wasted resources to lost time with a consistent, reliable workflow, which includes quality cover glass. Pre-cleaned Thermo Scientific™ Richard-Allan Scientific™ Cover Glasses are available in assorted sizes and thicknesses.

    Ordering Information:

    One oz. pack

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit