Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Thermo Scientific™ Richard-Allan Scientific™ Cover Glass Available on GSA/VA Contract for Federal Government customers only.
Reduces defect costs that range from wasted resources to lost time with a consistent, reliable workflow, which includes quality cover glass.
 View more versions of this product
Catalog No. SPN-102260
Description
Description
Reduces defect costs that range from wasted resources to lost time with a consistent, reliable workflow, which includes quality cover glass. Pre-cleaned Thermo Scientific™ Richard-Allan Scientific™ Cover Glasses are available in assorted sizes and thicknesses.
Ordering Information:
One oz. pack
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
22mm
0.13 to 0.17mm
60 x 22mm
60mm
Thickness no.1; 22 x 60mm