Learn More
Thermo Scientific™ Richard-Allan Scientific™ Dehydrant 100, 95, 80, 70 Available on GSA/VA Contract for Federal Government customers only.
Process and stain histological and cytological specimens with Thermo Scientific™ Richard-Allan Scientific™ Dehydrant 100, 95, 80, 70.
Manufacturer:  Thermo Scientific™ 6355
 View more versions of this product
Catalog No. 22-110-616
  Check Availability
Add to cart
Specifications
Description
Process and stain histological and cytological specimens with Thermo Scientific™ Richard-Allan Scientific™ Dehydrant 100, 95, 80, 70: a denatured ethyl alcohol reagent specifically designed for processing and staining of histological and cytological specimens.
  • Blend of ethyl, methyl and isopropyl alcohols
  • Can be used as a dye or stain solvent
  • Miscible with all clearing agents
Description & Specifications
Specifications
95%
55 gal.