Learn More
Thermo Scientific™ Richard-Allan Scientific™ Elastic Stain Available on GSA/VA Contract for Federal Government customers only.
Demonstrate the atrophy, thinning and loss of elastic tissue with Thermo Scientific™ Richard-Allan Scientific™ Elastic Stain.
$389.00 - $430.00
Products 2
Catalog Number Mfr. No. Type Price Quantity & Availability  
Catalog Number Mfr. No. Type Price Quantity & Availability  
87017 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
87017
Stain Kit
Each for $430.00
Add to cart
22-110-649
View Documents
Thermo Scientific™
88007
Alcoholic Hematoxylin Solution
Each for $389.00
Add to cart
Description
Description
Demonstrate the atrophy, thinning and loss of elastic tissue with Thermo Scientific™ Richard-Allan Scientific™ Elastic Stain.
  • Based on the Verhoeff Elastic technique
  • Also used for identifying the breaks, splitting and reduplication of elastic lamellae that occur in vascular diseases
  • Elastic fibers are stained blue-black to-black with red collagen and a yellow background
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.