Learn More
Thermo Scientific™ Richard-Allan Scientific™ Fast-Flex™ Processing Reagents
Improve the quality of processed tissue with these processing reagents, which can either be ordered with a xylene-based or a xylene-substitute-based clearant.
Manufacturer:  Thermo Scientific™ 9802
 View more versions of this product
Catalog No. 23-859-102
  Check Availability
Add to cart
Specifications
Description
Improve the quality of processed tissue with Thermo Scientific™ Richard-Allan Scientific™ Fast Flex™ Processing Reagents, which can either be ordered with a xylene-based or a xylene-substitute-based clearant.
  • Reduces Tissue processing time by 80%
  • Decreases the amount of reagents used up to 75%
  • Simplifies reagent management by eliminating the need to rotate reagents
  • Compatible with H&E, IHC, Special Stains and In situ
Includes:
One 5L bottle each: Fixative (Solution 1), Dehydrant (Solution 2) and Clearing Reagent (Solution 3).
Description & Specifications
Specifications
Fast Flex Kit 2, Xylene-free