Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Epredia™ Fast-Flex™ Processing Reagents
Improve the quality of processed tissue with these processing reagents, which can either be ordered with a xylene-based or a xylene-substitute-based clearant.
$634.00
Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
23-859-101
|
Epredia™
9801 |
Tissue Processing Reagent |
Each of 1 for $634.00
|
|
|||||
Description
- Reduces Tissue processing time by 80%
- Decreases the amount of reagents used up to 75%
- Simplifies reagent management by eliminating the need to rotate reagents
- Compatible with H&E, IHC, Special Stains and In situ
Includes:
One 5L bottle each: Fixative (Solution 1), Dehydrant (Solution 2) and Clearing Reagent (Solution 3).
Specifications
Tissue Processing Reagent |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title