Learn More
Thermo Scientific™ Richard-Allan Scientific™ Gram Stain (Tissue) Kit Available on GSA/VA Contract for Federal Government customers only.
Identify gram-positive and gram-negative bacteria in tissue sections with Thermo Scientific™ Richard-Allan Scientific™ Gram Stain (Tissue)
$222.50 - $610.00
Products 4
Catalog Number Mfr. No. Type Price Quantity & Availability  
Catalog Number Mfr. No. Type Price Quantity & Availability  
22-110-647 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
87018
Gram Stain (Tissue) Kit Each for $610.00
Add to cart
23-005-259
View Documents
Thermo Scientific™
88101
Crystal Violet Solution Each for $269.50
Add to cart
23-005-260
View Documents
Thermo Scientific™
88103
Safranin O Stain Solution Each for $248.50
Add to cart
23-005-261 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
88104
Decolorizing Solution Each for $222.50
Add to cart
Description
Description
Identify gram-positive and gram-negative bacteria in tissue sections with Thermo Scientific™ Richard-Allan Scientific™ Gram Stain (Tissue)
  • Gram-positive organisms stain blue to blue-black
  • Gram-negative organisms stain red
  • Tartrazine provides a yellow background stain
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.