Learn More
Epredia™ Richard-Allan Scientific™ Iron Stain, Nuclear Red Thiazin stain solution
Detect the presence of ferric iron (hemosiderin) in histological or hematological preparations with Epredia™ Richard-Allan Scientific™ Iron Stain.
Supplier: Epredia™ 88014
Description
- Useful in study of iron deficiency anemia, hemochromatosis and hemosiderosis
- Iron stains blue, nuclei stains pink-red, and cytoplasm stains pale pink
Specifications
Hydrochloric Acid Solution (20%) |