Thermo Scientific™ Richard-Allan Scientific™ Mucolytic Agent
Easily identify specimens through the tint of Thermo Scientific™ Richard-Allan Scientific™ Mucolytic Agent.
Manufacturer: Thermo Scientific™ 76166
 View more versions of this product
Catalog No. 22-050-216
  Check Availability
Add to Cart
    Specifications
    Product Type 16 oz.
    Type 16 oz.
    Easily identify specimens through the tint of Thermo Scientific™ Richard-Allan Scientific™ Mucolytic Agent.
    • Preserve and liquefy mucoid cytology specimens to optimize diagnostic results
    • Maintain cell morphology, size, and staining characteristics of fresh and unfixed cytology specimens
    • Store specimens indefinitely with no adverse effects
  • Description & Specifications
Related Products
Product Content Feedback