Learn More
Thermo Scientific™ Richard-Allan Scientific™ Mucolytic Agent Available on GSA/VA Contract for Federal Government customers only.
Easily identify specimens through the tint of Thermo Scientific™ Richard-Allan Scientific™ Mucolytic Agent.
Manufacturer:  Thermo Scientific™ 76167
 View more versions of this product
Catalog No. 22-050-299
  Check Availability
Add to cart
Specifications
Description
Easily identify specimens through the tint of Thermo Scientific™ Richard-Allan Scientific™ Mucolytic Agent.
  • Preserve and liquefy mucoid cytology specimens to optimize diagnostic results
  • Maintain cell morphology, size, and staining characteristics of fresh and unfixed cytology specimens
  • Store specimens indefinitely with no adverse effects
Description & Specifications
Specifications
1 gal.