Learn More
Thermo Scientific™ Richard-Allan Scientific™ Mucolytic Agent
Easily identify specimens through the tint of Thermo Scientific™ Richard-Allan Scientific™ Mucolytic Agent.
Specifications
Type Mucolytic Agent
View More Specs
Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
22-050-216
View Documents
Thermo Scientific™
76166
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Due to product restrictions, please to purchase or view availability for this product.
N/A
Description
Description
Easily identify specimens through the tint of Thermo Scientific™ Richard-Allan Scientific™ Mucolytic Agent.
  • Preserve and liquefy mucoid cytology specimens to optimize diagnostic results
  • Maintain cell morphology, size, and staining characteristics of fresh and unfixed cytology specimens
  • Store specimens indefinitely with no adverse effects
Specifications
Specifications
Mucolytic Agent