Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Thermo Scientific™ Richard-Allan Scientific™ Periodic Acid-Schiff (PAS) Available on GSA/VA Contract for Federal Government customers only.
Demonstrate the patterns and presence of specific carbohydrates with Thermo Scientific™ Richard-Allan Scientific™ Periodic Acid-Schiff (PAS) Solution
$129.00 - $476.50
Products 2
Catalog Number Mfr. No. Type Price Quantity & Availability  
Catalog Number Mfr. No. Type Price Quantity & Availability  
22-110-645 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
87007
Periodic Acid-Schiff (PAS) Kit Each for $476.50
N/A
88-018
View Documents
Thermo Scientific™
88018
Hematoxylin I Each for $129.00
N/A
Description
Description
Demonstrate the patterns and presence of specific carbohydrates with Thermo Scientific™ Richard-Allan Scientific™ Periodic Acid-Schiff (PAS) Solution
  • Glycogen, mucin, fungal walls and some basement membranes stain rose to magenta
  • Nuclei stain blue
  • Kit includes: Hematoxylin I, periodic acid solution (0.5%), and Schiff reagent
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.