Learn More
Epredia™ Reticulin Silver Stain Kit
Identify and visualize reticular fibers in tissue sections with the Reticulin Silver Stain, a Gordon and Sweet's modification.
Supplier: Epredia™ 87025
This item is not returnable.
View return policy
Description
- Working ammoniacal silver solution selectively impregnates and reacts with reticular fibers through multiple reactions, including oxidation and reduction
- Nuclear Fast Red Stain Solution provides red background stain
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov