Learn More
Thermo Scientific™ Richard-Allan Scientific™ Reticulin Silver Available on GSA/VA Contract for Federal Government customers only.
Identify and visualize reticular fibers in tissue sections with the Reticulin Silver Stain, a Gordon and Sweet's modification.
$1232.00
Products 1
Catalog Number Mfr. No. Type Price Quantity & Availability  
Catalog Number Mfr. No. Type Price Quantity & Availability  
23-900-666 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
87025
Stain Kit Each for $1,232.00
Add to cart
Description
Description
Identify and visualize reticular fibers in tissue sections with the Thermo Scientific™ Richard-Allan Scientific™ Reticulin Silver Stain, a Gordon and Sweet's modification.
  • Working ammoniacal silver solution selectively impregnates and reacts with reticular fibers through multiple reactions, including oxidation and reduction
  • Nuclear Fast Red Stain Solution provides red background stain
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.