Thermo Scientific™ Richard-Allan Scientific™ Reusable Steel Knives Available on GSA/VA Contract for Federal Government customers only.
Use Thermo Scientific™ Richard-Allan Scientific™ Reusable Steel Knives with microtomes or cryostats when the technical application dictates that requirement.
Manufacturer: Thermo Scientific™ 152010
 View more versions of this product
Catalog No. 14-070-24
  Check Availability
Add to Cart
    Specifications
    Product Line Richard-Allan Scientific™
    Type Steel Knives
    Profile C
    Item Description Knife-12cm, C Profile Steel
    Description Knife
    Disposable No
    Length (Metric) 12cm
    Use Thermo Scientific™ Richard-Allan Scientific™ Reusable Steel Knives with microtomes or cryostats when the technical application dictates that requirement.
    • Permanent knives can be resharpened on an appropriate knife sharpener for extended use
    • Appropriate knife case(s) should be ordered with the knife or knives, each case holds two knives
    Multiple Styles Available:
    • C-profile knives are used for standard paraffin and cryo techniques
    • D-profile knives are used for sectioning of harder materials that require greater knife stability
    • Tungsten carbide knives are used for strength and durability in sectioning of very hard samples or embedded resin
  • Description & Specifications
Related Products
Product Content Feedback