Thermo Scientific™ Richard-Allan Scientific™ Reusable Steel Knives
Use Thermo Scientific™ Richard-Allan Scientific™ Reusable Steel Knives with microtomes or cryostats when the technical application dictates that requirement.
Manufacturer: Thermo Scientific™ 152120
 View more versions of this product
Catalog No. 15-212-0
  Check Availability
Add to Cart
    Specifications
    Product Line Richard-Allan Scientific™
    Type Steel Knives
    Profile D
    Item Description D-Profile Tungsten Carbide Tip; 16cm
    Description Tungsten Carbide Tip
    Length (Metric) 16cm
    Use Thermo Scientific™ Richard-Allan Scientific™ Reusable Steel Knives with microtomes or cryostats when the technical application dictates that requirement.
    • Permanent knives can be resharpened on an appropriate knife sharpener for extended use
    • Appropriate knife case(s) should be ordered with the knife or knives, each case holds two knives
    Multiple Styles Available:
    • C-profile knives are used for standard paraffin and cryo techniques
    • D-profile knives are used for sectioning of harder materials that require greater knife stability
    • Tungsten carbide knives are used for strength and durability in sectioning of very hard samples or embedded resin
  • Description & Specifications
Related Products
Product Content Feedback