Learn More
Thermo Scientific™ Richard-Allan Scientific™ Reusable Steel Knives
Use Thermo Scientific™ Richard-Allan Scientific™ Reusable Steel Knives with microtomes or cryostats when the technical application dictates that requirement.
Manufacturer:  Thermo Scientific™ 152080
 View more versions of this product
Catalog No. 15-208-0
  Check Availability
Add to cart
Specifications
Description
Use Thermo Scientific™ Richard-Allan Scientific™ Reusable Steel Knives with microtomes or cryostats when the technical application dictates that requirement.
  • Permanent knives can be resharpened on an appropriate knife sharpener for extended use
  • Appropriate knife case(s) should be ordered with the knife or knives, each case holds two knives
Multiple Styles Available:
  • C-profile knives are used for standard paraffin and cryo techniques
  • D-profile knives are used for sectioning of harder materials that require greater knife stability
  • Tungsten carbide knives are used for strength and durability in sectioning of very hard samples or embedded resin
Description & Specifications
Specifications
Richard-Allan Scientific™
Steel Knives
22cm
Knife
D
No
Steel D-Profile Knives; 22cm