Learn More
Thermo Scientific™ Richard-Allan Scientific™ Reusable Steel Knives
Use Thermo Scientific™ Richard-Allan Scientific™ Reusable Steel Knives with microtomes or cryostats when the technical application dictates that requirement.
Specifications
Product Line Richard-Allan Scientific™
Type Steel Knives
View More Specs
Products 3
Catalog Number Mfr. No. Description Profile Length (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Description Profile Length (Metric) Price Quantity & Availability  
14-070-24
View Documents
Thermo Scientific™
152010
Knife C 12cm This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
15-227-0
View Documents
Thermo Scientific™
152270
Knife C 18.5cm This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
152460M
View Documents
Thermo Scientific™
152460M
Solid Tungsten D 16cm This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
Use Thermo Scientific™ Richard-Allan Scientific™ Reusable Steel Knives with microtomes or cryostats when the technical application dictates that requirement.
  • Permanent knives can be resharpened on an appropriate knife sharpener for extended use
  • Appropriate knife case(s) should be ordered with the knife or knives, each case holds two knives
Multiple Styles Available:
  • C-profile knives are used for standard paraffin and cryo techniques
  • D-profile knives are used for sectioning of harder materials that require greater knife stability
  • Tungsten carbide knives are used for strength and durability in sectioning of very hard samples or embedded resin
Specifications
Specifications
Richard-Allan Scientific™
Steel Knives