Learn More
Thermo Scientific™ Richard-Allan Scientific™ Signature Series Clear-Rite™ 3
Render complete deparaffinization and clearing during the staining process with Thermo Scientific™ Richard-Allan Scientific™ Signature Series Clear-Rite™ 3.
Manufacturer:  Thermo Scientific™ 6955
 View more versions of this product
Catalog No. 22-110-619
  Check Availability
Add to cart
Specifications
Description
Render complete deparaffinization and clearing during the staining process with Thermo Scientific™ Richard-Allan Scientific™ Signature Series Clear-Rite™ 3, a unique blend of isoparaffinic aliphatic hydrocarbons.
  • Very low odor; Benzene-free
  • Use in all tissue processors and automatic stainers
  • Does not produce hard or brittle specimens, even after prolonged exposure
Description & Specifications
Specifications
Clearing Agent; Drum
Clearing Agent; Drum
55 gal.