Thermo Scientific™ Richard-Allan Scientific™ Signature Series Flex™ 100, 95, 80
Achieve brilliant cytoplasmic staining and complete dehydration with Thermo Scientific™ Richard-Allan Scientific™ Signature Series Flex™ 100, 95, 80.
Manufacturer: Thermo Scientific™ 8105
 View more versions of this product
Catalog No. 23-900-650
  Check Availability
Add to Cart
    Specifications
    Product Type Flex 100
    Quantity 5 gal.,5 gal.
    Type Flex 100
    Item Description Flex 100; 5 Gal. (18.9L) Metal Pail
    Achieve brilliant cytoplasmic staining and complete dehydration with Thermo Scientific™ Richard-Allan Scientific™ Signature Series Flex™ 100, 95, 80.
    • Patented formulation of methyl and isopropyl alcohol minimizes over-dehydration in smaller biopsies
    • Excellent for lipid extraction allowing for increased paraffin infiltration
    • May be used as a solvent for special stains
  • Description & Specifications
Related Products
Product Content Feedback