Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Richard-Allan Scientific™ Signature Series Hematoxylin 7212

Yield results which exceed those of Gill or Harris Hematoxylins with this uniquely formulated product.

Manufacturer:  Thermo Scientific™ 7212

 View more versions of this product

Catalog No. 22-050-306


Add to cart

Description

Description

Yield results that exceed those of Gill or Harris Hematoxylins with the Thermo Scientific™ Richard-Allan Scientific™ Signature Series Hematoxylin 7212, a uniquely formulated product.

  • No filtration required
  • Stains up to 2500 slides per pint
  • Crisp, well-defined nuclear detail
  • Slight affinity for acid mucopolysaccharide (mucin) staining

Available in 5 liter packaging

  • Reduces plastic by 90%
  • Increases stain stability
  • Reduces messy stain spills
  • Saves 35% storage space versus traditional packaging
Specifications

Specifications

Hematoxylin
1 pt.
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit