Thermo Scientific™ Richard-Allan Scientific™ Specimen Transportation System (STS™ ) Available on GSA/VA Contract for Federal Government customers only.

Transport, store and retrieve pathology specimens in a safe and cost effecetive manner with the Thermo Scientific™ Richard-Allan Scientific™ Specimen Transportation System (STS™).

Manufacturer: Thermo Scientific™ 599601

 View more versions of this product

Catalog No. 22-110-664

  Check Availability
Add to Cart

    Specifications

    Description Prefilled Container, 10% NBF
    No. per Case 12/CS
    Sterile No
    Material Polypropylene
    Cap Type Screw
    Capacity (Metric) 960mL

    Transport, store and retrieve pathology specimens in a safe and cost effective manner with the Thermo Scientific™ Richard-Allan Scientific™ Specimen Transportation System (STS™). This system includes containers pre-filled with the highest quality 10% Neutral Buffered Formalin (NBF) and Zinc Formalin and features a unique Signature Series tray packing for easy distribution and transport. 

    Closure System

    • Patented Thermo Scientific™ Bio-Tite™ closure system is dual-threaded, making it virtually impossible to cross-thread and minimizing leakage
    Pre-Filled Containers
    • Nonsterile containers marked with a lot code and expiration date
    • Comply with all updated federal and state OSHA requirement
    • Labeled with highly visible, bright yellow labels with space for patient information for instant identification to minimize health risks

    Ordering Information

    • Clear polystyrene containers have screw-top lids
    • Opaque polyethylene containers have snap-top lids
    • Dry containers available separately

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit