Thermo Scientific™ Richard-Allan Scientific™ Super U-Rite™ and Bond-Rite™ Slides Available on GSA/VA Contract for Federal Government customers only.
Optimize tissue adhesion with Thermo Scientific™ Richard-Allan Scientific™ Super Up-Rite™ and Bond-Rite™ Slides, manufactured with the finest optical quality white glass.
Manufacturer: Thermo Scientific™ 4431
 View more versions of this product
Catalog No. 22-046-501
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »
    Specifications
    Description Super Up-Rite
    Color Blue
    Item Description Super Up-Rite*; Blue; 1440/Cs.
    Optimize tissue adhesion with Thermo Scientific™ Richard-Allan Scientific™ Super Up-Rite™ and Bond-Rite™ Slides, manufactured with the finest optical quality white glass.
    • Slides are available in five colors for easy identification
    • Bond-Rite slides are specially treated to optimize adhesion of the tissue to the slide
  • Description & Specifications
Related Products
Product Content Feedback