Learn More
Thermo Scientific™ Richard-Allan Scientific™ Super U-Rite™ and Bond-Rite™ Slides
Optimize tissue adhesion with Thermo Scientific™ Richard-Allan Scientific™ Super Up-Rite™ and Bond-Rite™ Slides, manufactured with the finest optical quality white glass.
Manufacturer:  Thermo Scientific™ 4432
 View more versions of this product
Catalog No. 22-046-502
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications
Description
Optimize tissue adhesion with Thermo Scientific™ Richard-Allan Scientific™ Super Up-Rite™ and Bond-Rite™ Slides, manufactured with the finest optical quality white glass.
  • Slides are available in five colors for easy identification
  • Bond-Rite slides are specially treated to optimize adhesion of the tissue to the slide
Description & Specifications
Specifications
Super Up-Rite
Green
Super Up-Rite*; Green; 1440/Cs.